Dihexyl ether ≥98.0%

Supplier: TCI America

H0138-25ML H0138-500ML
TCH0138-025MLEA 75.57 CAD
TCH0138-025ML TCH0138-500ML
Dihexyl ether ≥98.0%
Dihexyl ether

CAS Number: 112-58-3
MDL Number: MFCD00009525
Molecular Formula: C12H26O
Molecular Weight: 186.34
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 226
Melting point (°C): -43
Flash Point (°C): 77
Specific Gravity (20/20): 0.79


Other Product Names / Synonyms: Dihexyl Ether

Formula: C₁₂H₂₆O
Boiling Pt: 226 °C
Melting Pt: –43 °C
Density: 0.79 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00009525
CAS Number: 112-58-3
EINECS: 203-987-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc35ee2bdf89c6551fe2075abd898f31ec850f20&vid=cc35ee2bdf89c6551fe2075abd898f31ec850f20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR