Sodium hexanoate ≥99.0%

Supplier: TCI America

Synonyms: N-caproicacidsodiumsalt, Sodium Hexanoate

H0112-500G H0112-25G
TCH0112-500GEA 578.28 CAD
TCH0112-500G TCH0112-025G
Sodium hexanoate ≥99.0%
Sodium hexanoate

CAS Number: 10051-44-2
MDL Number: MFCD00059056
Molecular Formula: C6H12O2
Molecular Weight: 138.14
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Hexanoic Acid Sodium Salt

Formula: C₆H₁₁NaO₂
MDL Number: MFCD00059056
CAS Number: 10051-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=704f56f86822f98289db335454a68fb762a23fa4&vid=704f56f86822f98289db335454a68fb762a23fa4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR