Pentyl hexanoate ≥98.0%
Supplier: TCI America
H0106-25ML
H0106-500ML
TCH0106-025MLEA
42.66
CAD
TCH0106-025ML
TCH0106-500ML
Pentyl hexanoate ≥98.0%
Pentyl hexanoate
CAS Number: 540-07-8
MDL Number: MFCD00027281
Molecular Formula: C11H22O2
Molecular Weight: 186.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 226
Flash Point (°C): 91
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: Hexanoic Acid Amyl Ester, Pentyl Hexanoate, Hexanoic Acid Pentyl Ester
Formula:
C₁₁H₂₂O₂ Boiling Pt: 226 °C Melting Pt: –47 °C Density: 0.87 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD00027281 CAS Number: 540-07-8 EINECS: 208-732-4 Merck Index: 13,00609 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...