Hexamethylphosphoramide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0095-500G H0095-25G H0095-100G
TCH0095-500GEA 266.28 CAD
TCH0095-500G TCH0095-025G TCH0095-100G
Hexamethylphosphoramide ≥98.0% (by GC, titration analysis)
Hexamethylphosphoramide

CAS Number: 680-31-9
MDL Number: MFCD00008303
Molecular Formula: C6H18N3OP
Molecular Weight: 179.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 232
Flash Point (°C): 132
Freezing point (°C): 6
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: HMPA

Formula: C₆H₁₈N₃OP
Boiling Pt: 232 °C
Density: 1.03 (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00008303
CAS Number: 680-31-9
EINECS: 211-653-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=718f1b5f37c7d822df3c9e0d7d79be0409423401&vid=718f1b5f37c7d822df3c9e0d7d79be0409423401&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR