8-Nonenyl acetate ≥98.0%
Supplier: TCI America
A1384-25ML
A1384-5ML
TCA1384-25MLEA
507.18
CAD
TCA1384-25ML
TCA1384-005ML
8-Nonenyl acetate ≥98.0%
8-Nonenyl acetate
CAS Number: 13038-22-7
MDL Number: MFCD00671492
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Specific Gravity (20/20): 0.88
Other Product Names / Synonyms: 9-Acetoxy-1-nonene, Acetic Acid 8-Nonenyl Ester
Formula:
C₁₁H₂₀O₂ Boiling Pt: 155 °C Density: 0.88 (20 °C) |
MDL Number:
MFCD00671492 CAS Number: 13038-22-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...