Palmitoleic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (Z)-Hexadec-9-enoic acid

H0072-1G H0072-100MG
TCH0072-001GEA 395 CAD
TCH0072-001G TCH0072-100MG
Palmitoleic acid ≥98.0% (by GC, titration analysis)
Palmitoleic acid

CAS Number: 373-49-9
MDL Number: MFCD00004437
Molecular Formula: C16H30O2
Molecular Weight: 254.41
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 175
Melting point (°C): 1
Flash Point (°C): 62
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Palmitoleic Acid

Formula: C₁₆H₃₀O₂
MW: 254.41 g/mol
Boiling Pt: 175 °C
Melting Pt: 1 °C
Density: 0.90 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00004437
CAS Number: 373-49-9
EINECS: 206-765-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b234331a3a6bd412ec16643e90d2ec9b43e8494&vid=8b234331a3a6bd412ec16643e90d2ec9b43e8494&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR