Trisodium-N-(2-hydroxyethyl)ethylenediamine-N,N',N'-triacetate dihydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic Acid Trisodium Salt

H0047-500G H0047-25G
TCH0047-500GEA 190.83 CAD
TCH0047-500G TCH0047-025G
Trisodium-N-(2-hydroxyethyl)ethylenediamine-N,N',N'-triacetate dihydrate ≥98.0% (by titrimetric analysis)
Trisodium-N-(2-hydroxyethyl)ethylenediamine-N,N',N'-triacetate dihydrate

CAS Number: 139-89-9
MDL Number: MFCD00043341
Molecular Formula: C10H15N2Na3O7·2H2O
Molecular Weight: 344.21(as Anhydrous)
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 288


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: N-Carboxymethyl-N'-(2-hydroxyethyl)-N,N'-ethylenediglycine Trisodium Salt, Versenol Trisodium Salt, N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic Acid Trisodium Salt, HEDTA 3Na

Formula: C₁₀H₁₅N₂Na₃O₇. 2H₂O
MW: 344.21 g/mol (anhy)
Melting Pt: 288 °C
MDL Number: MFCD00043341
CAS Number: 139-89-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec24b6c68b1f64e8a3b7af8dfd3476e142bb1689&vid=4d2a5dacbfe2befd5eedeb441617ed70695c0abe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR