Glucan (from black yeast)
Supplier: TCI America
G0331-5G
G0331-1G
TCG0331-5GEA
666.76
CAD
TCG0331-5G
TCG0331-1G
Glucan (from black yeast)
Glucan
CAS Number: 9012-72-0
MDL Number: MFCD00466924
Form: Crystal
Color: Very Pale Yellow
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides
Formula:
(C₆H₁₀O₅)n |
MDL Number:
MFCD00466924 CAS Number: 9012-72-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Beta-1,3-1,6-Glucan | min. 80 % |
Drying loss | max. 8.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...