(S)-N-Glycidylphthalimide ≥98.0%
Supplier: TCI America
G0328-5G
G0328-25G
TCG0328-5GEA
71.1
CAD
TCG0328-5G
TCG0328-25G
(S)-N-Glycidylphthalimide ≥98.0%
(S)-N-Glycidylphthalimide
CAS Number: 161596-47-0
MDL Number: MFCD04973350
Molecular Formula: C11H9NO3
Molecular Weight: 203.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 102
Specific rotation [a]20/D: 10 deg (C=2.2, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-N-(2,3-Epoxypropyl)phthalimide
Formula:
C₁₁H₉NO₃ Melting Pt: 102 °C |
MDL Number:
MFCD04973350 CAS Number: 161596-47-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...