(R)-N-Glycidylphthalimide ≥99.0%
Supplier: TCI America
G0327-5G
G0327-25G
TCG0327-5GEA
96.38
CAD
TCG0327-5G
TCG0327-25G
(R)-N-Glycidylphthalimide ≥99.0%
(R)-N-Glycidylphthalimide
CAS Number: 181140-34-1
MDL Number: MFCD04973349
Molecular Formula: C11H9NO3
Molecular Weight: 203.20
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 101
Specific rotation [a]20/D: -10 deg (C=2.2, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-N-(2,3-Epoxypropyl)phthalimide
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...