Galactinol hydrate ≥99.0% (by HPLC)

Supplier: TCI America

G0298-100MG G0298-1G
TCG0298-100MGEA 189.6 CAD
TCG0298-100MG TCG0298-1G
Galactinol hydrate ≥99.0% (by HPLC)
Galactinol hydrate

CAS Number: 3687-64-7
Molecular Formula: C12H22O11
Molecular Weight: 342.30
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 221
Specific rotation [a]20/D: 132 deg (C=1, H2O)


Product Categories: Bioscience, Disaccharides
Other Product Names / Synonyms: 1-O-alpha-D-Galactopyranosyl-L-myo-inositol

Formula: C₁₂H₂₂O₁₁
Melting Pt: 221 °C
CAS Number: 3687-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d3bdf303867da340505bc90452804c3d51ca05a&vid=6d3bdf303867da340505bc90452804c3d51ca05a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR