(S)-Glycidyl trityl ether ≥98.0%

Supplier: TCI America

Synonyms: (S)-Glycidyl Triphenylmethyl Ether, (S)-2-(Triphenylmethoxymethyl)oxirane, (S)-2-((Trityloxy)methyl)oxirane

G0285-5G G0285-25G
TCG0285-5GEA 109.02 CAD
TCG0285-5G TCG0285-25G
(S)-Glycidyl trityl ether ≥98.0%
(S)-Glycidyl trityl ether

CAS Number: 129940-50-7
MDL Number: MFCD00273373
Molecular Formula: C22H20O2
Molecular Weight: 316.40
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 101
Flash Point (°C): 110
Specific rotation [a]20/D: -11 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-Glycidyl Triphenylmethyl Ether, (S)-2-(Triphenylmethoxymethyl)oxirane

Formula: C₂₂H₂₀O₂
MW: 316.40 g/mol
Melting Pt: 101 °C
Flash Pt: 110 °C
MDL Number: MFCD00273373
CAS Number: 129940-50-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313ed0b077f6d67aef03a485b62bec6ef11aad68&vid=313ed0b077f6d67aef03a485b62bec6ef11aad68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR