(R)-Glycidyl trityl ether ≥98.0%

Supplier: TCI America

Synonyms: (R)-2-(Triphenylmethoxymethyl)oxirane, (R)-(+)-Glycidyl trityl ether, (R)-Glycidyl Triphenylmethyl Ether

G0284-25G G0284-5G
TCG0284-25GEA 900.6 CAD
TCG0284-25G TCG0284-5G
(R)-Glycidyl trityl ether ≥98.0%
(R)-Glycidyl trityl ether

CAS Number: 65291-30-7
MDL Number: MFCD00273368
Molecular Formula: C22H20O2
Molecular Weight: 316.40
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 102
Flash Point (°C): 110
Specific rotation [a]20/D: 11 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-Glycidyl Triphenylmethyl Ether, (R)-2-(Triphenylmethoxymethyl)oxirane

Formula: C₂₂H₂₀O₂
MW: 316.40 g/mol
Melting Pt: 102 °C
Flash Pt: 110 °C
MDL Number: MFCD00273368
CAS Number: 65291-30-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1c6cfbf08b7a72256e98f099ec13e529a48477e&vid=c1c6cfbf08b7a72256e98f099ec13e529a48477e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR