D-(-)-Glutamine ≥98.0%

Supplier: TCI America

Synonyms: H-D-Gln-OH, (R)-2,5-Diamino-5-oxopentanoic acid, (R)-(-)-Glutamine

G0278-1G G0278-5G
TCG0278-1GEA 48.21 CAD
TCG0278-1G TCG0278-5G
D-(-)-Glutamine ≥98.0%
D(-)-Glutamine

CAS Number: 5959-95-5
MDL Number: MFCD00065607
Molecular Formula: C5H10N2O3
Molecular Weight: 146.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -6.8 deg (C=4, H2O)


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Gln-OH

Formula: C₅H₁₀N₂O₃
MW: 146.15
MDL Number: MFCD00065607
CAS Number: 5959-95-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3f046fbc0f8ca30b81615db5c5a305ad0d16941&vid=f3f046fbc0f8ca30b81615db5c5a305ad0d16941&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR