3-Glycidyloxypropyl(dimethoxy)methylsilane ≥96.0%

Supplier: TCI America

G0261-500G G0261-25G
TCG0261-500GEA 1232.4 CAD
TCG0261-500G TCG0261-025G
3-Glycidyloxypropyl(dimethoxy)methylsilane ≥96.0%
3-Glycidyloxypropyl(dimethoxy)methylsilane

CAS Number: 65799-47-5
MDL Number: MFCD00271003
Molecular Formula: C9H20O4Si
Molecular Weight: 220.34
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 132
Specific Gravity (20/20): 1.02


Product Categories: Materials Chemistry, Silane Coupling Agents, Monomers
Other Product Names / Synonyms: 3-[Dimethoxy(methyl)silyl]propyl Glycidyl Ether

Formula: C₉H₂₀O₄Si
MDL Number: MFCD00271003
CAS Number: 65799-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ec15dea59e37560851d8ff5f49aa3144ee6037c&vid=9ec15dea59e37560851d8ff5f49aa3144ee6037c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR