2-[2-(2-Azidoethoxy)ethoxy]ethyl-2,3,4,6-tetra-O-acetyl-D-galactopyranoside ≥95.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 2-[2-(2-Azidoethoxy)ethoxy]ethyl 2,3,4,6-Tetra-O-acetyl-D-galactopyranoside

G0257-5G G0257-1G
TCG0257-5GEA 1079.3 CAD
TCG0257-5G TCG0257-001G
2-[2-(2-Azidoethoxy)ethoxy]ethyl-2,3,4,6-tetra-O-acetyl-D-galactopyranoside ≥95.0% (by total nitrogen basis)
2-[2-(2-Azidoethoxy)ethoxy]ethyl-2,3,4,6-tetra-O-acetyl-D-galactopyranoside

CAS Number: 381716-33-2
MDL Number: MFCD00191441
Molecular Formula: C20H31N3O12
Molecular Weight: 505.48
Purity/Analysis Method: >95.0% (N)
Form: Clear Liquid

Formula: C₂₀H₃₁N₃O₁₂
MW: 505.48 g/mol
MDL Number: MFCD00191441
CAS Number: 381716-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8117e399b7b14509fbd0fda6c45aa882f5c66079&vid=8117e399b7b14509fbd0fda6c45aa882f5c66079&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR