4-Guanidinobenzoic acid methanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

G0248-5G G0248-25G
TCG0248-005GEA 45.82 CAD
TCG0248-005G TCG0248-025G
4-Guanidinobenzoic acid methanesulfonate ≥98.0% (by titrimetric analysis)
4-Guanidinobenzoic acid methanesulfonate

CAS Number: 148720-07-4
MDL Number: MFCD00191442
Molecular Formula: C8H9N3O2
Molecular Weight: 275.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 228

Formula: C₈H₉N₃O₂
MW: 275.28 g/mol
Melting Pt: 228 °C
MDL Number: MFCD00191442
CAS Number: 148720-07-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5708c9824a8f7851ce96bcf9f85bbf32a0c0a6f&vid=b5708c9824a8f7851ce96bcf9f85bbf32a0c0a6f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR