Glycerol formal (mixture of 1,3-dioxan-5-ol and 4-hydroxymethyldioxolane) ≥98.0%

Supplier: TCI America

G0244-25ML G0244-500ML
TCG0244-025MLEA 53.72 CAD
TCG0244-025ML TCG0244-500ML
Glycerol formal (mixture of 1,3-dioxan-5-ol and 4-hydroxymethyldioxolane) ≥98.0%
Glycerol formal (mixture of 1,3-dioxan-5-ol and 4-hydroxymethyldioxolane)

CAS Number: 99569-11-6
MDL Number: MFCD00003218
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 93
Specific Gravity (20/20): 1.22

Formula: C₄H₈O₃
Boiling Pt: 97 °C
Density: 1.22 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00003218
CAS Number: 99569-11-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e60eeced9d876b3654b9e4e2ff9736cb7b25636a&vid=e60eeced9d876b3654b9e4e2ff9736cb7b25636a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR