L-(+)-Gulonic acid-γ-lactone ≥98.0%
Supplier: TCI America
G0235-5G
G0235-25G
TCG0235-5GEA
123.81
CAD
TCG0235-5G
TCG0235-025G
L-(+)-Gulonic acid-γ-lactone ≥98.0%
L-(+)-Gulonic acid-gamma-lactone
CAS Number: 1128-23-0
MDL Number: MFCD00064331
Molecular Formula: C6H10O6
Molecular Weight: 178.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 188
Specific rotation [a]20/D: 56 deg (C=2, H2O)
Product Categories: Bioscience, Sugar Acids, Gulose
Other Product Names / Synonyms: L-Gulonolactone
Formula:
C₆H₁₀O₆ Melting Pt: 188 °C |
MDL Number:
MFCD00064331 CAS Number: 1128-23-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...