DL-Glyceraldehyde diethyl acetal ≥95.0%
Supplier: TCI America
G0216-25G
G0216-5G
TCG0216-025GEA
1338.26
CAD
TCG0216-025G
TCG0216-005G
DL-Glyceraldehyde diethyl acetal ≥95.0%
DL-Glyceraldehyde diethyl acetal
CAS Number: 10487-05-5
MDL Number: MFCD00059599
Molecular Formula: C7H16O4
Molecular Weight: 164.20
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Specific Gravity (20/20): 1.06
Formula:
C₇H₁₆O₄ Boiling Pt: 136 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD00059599 CAS Number: 10487-05-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...