Glycine zinc salt monohydrate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
G0215-5G
G0215-25G
TCG0215-5GEA
151.08
CAD
TCG0215-5G
TCG0215-025G
Glycine zinc salt monohydrate ≥97.0% (by titrimetric analysis)
Glycine zinc salt monohydrate
[for Protein Research]
CAS Number: 14281-83-5
MDL Number: MFCD00070561
Molecular Formula: C2H5NO2
Molecular Weight: 213.50
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: Zinc Glycinate
Formula:
C₂H₅NO₂ |
MDL Number:
MFCD00070561 CAS Number: 14281-83-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...