Sodium glucoheptonate dihydrate ≥98.0%
Supplier: TCI America
Synonyms:
Glucosecarboxylic Acid Sodium Salt, Glucoheptonic Acid Sodium Salt
G0214-25G
G0214-500G
TCG0214-025GEA
61.15
CAD
TCG0214-025G
TCG0214-500G
Sodium glucoheptonate dihydrate ≥98.0%
Sodium glucoheptonate dihydrate
CAS Number: 31138-65-5
MDL Number: MFCD00013425
Molecular Formula: C7H14O8
Molecular Weight: 248.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 5.5 deg (C=10, H2O)
Product Categories: Bioscience, Sugar Acids, Sugars (Others)
Other Product Names / Synonyms: Glucoheptonic Acid Sodium Salt, Glucosecarboxylic Acid Sodium Salt
Formula:
C₇H₁₄O₈ MW: 248.16 g/mol |
MDL Number:
MFCD00013425 CAS Number: 31138-65-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...