Tristearin (glycerol tristearate) ≥80.0%

Supplier: TCI America

G0212-25G G0212-250G
TCG0212-025GEA 157.94 CAD
TCG0212-025G TCG0212-250G
Tristearin (glycerol tristearate) ≥80.0%
Tristearin (Glycerol tristearate)

CAS Number: 555-43-1
MDL Number: MFCD00036230
Molecular Formula: C57H110O6
Molecular Weight: 891.50
Purity/Analysis Method: >80.0% (GC)
Form: Crystal
Melting point (°C): 73


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Tristearate

Formula: C₅₇H₁₁₀O₆
Melting Pt: 73 °C
MDL Number: MFCD00036230
CAS Number: 555-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=337c43d5901254c397bbadb82b4f6d27b35932a5&vid=337c43d5901254c397bbadb82b4f6d27b35932a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR