Guanidinium hydrochloride ≥99.0% for synthesis

Supplier: TCI America
Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

G0197-25G G0197-500G G0197-100G
TCG0197-025GEA 29.55 CAD
TCG0197-025G TCG0197-500G TCG0197-100G
Guanidinium hydrochloride ≥99.0% for synthesis
Guanidine hydrochloride

[for Protein Research]
CAS Number: 50-01-1
MDL Number: MFCD00013026
Molecular Formula: CH5N3
Molecular Weight: 95.53
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 187

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52e4dd9f0a4b1455d358721a1fb2ac60050f650c&vid=2fe27eedee6de0a811023e1e664a52fd3375cc70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR