3-Allyloxy-1,2-propanediol ≥99.0%
Supplier: TCI America
G0189-25G
G0189-500G
TCG0189-025GEA
43.37
CAD
TCG0189-025G
TCG0189-500G
3-Allyloxy-1,2-propanediol ≥99.0%
3-Allyloxy-1,2-propanediol
CAS Number: 123-34-2
MDL Number: MFCD00004721
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 134
Specific Gravity (20/20): 1.07
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Glycerol alpha-Monoallyl Ether
Formula:
C₆H₁₂O₃ Boiling Pt: 142 °C Density: 1.07 (20 °C) Flash Pt: 134 °C |
MDL Number:
MFCD00004721 CAS Number: 123-34-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...