Glycyl-L-leucine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

G0181-10G G0181-1G G0181-5G
TCG0181-010GEA 530.88 CAD
TCG0181-010G TCG0181-001G TCG0181-5G
Glycyl-L-leucine ≥99.0% (by titrimetric analysis)
Glycyl-L-leucine

CAS Number: 869-19-2
MDL Number: MFCD00008127
Molecular Formula: C8H16N2O3
Molecular Weight: 188.23
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 235
Specific rotation [a]20/D: -35 deg (C=2, H2O)


Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Leu-OH

Formula: C₈H₁₆N₂O₃
Melting Pt: 235 °C
MDL Number: MFCD00008127
CAS Number: 869-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28ff84354d47ccccce335d9272c45c47bc8e6585&vid=28ff84354d47ccccce335d9272c45c47bc8e6585&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR