Guanine sulfate dihydrate ≥98.0%

Supplier: TCI America

Synonyms: 2-Aminohypoxanthine Sulphate, 2-Aminohypoxanthine Sulfate

G0168-1G G0168-25G
TCG0168-001GEA 42.66 CAD
TCG0168-001G TCG0168-025G
Guanine sulfate dihydrate ≥98.0%
Guanine sulfate dihydrate

CAS Number: 10333-92-3
MDL Number: MFCD00213671
Molecular Formula: C5H5N5O
Molecular Weight: 400.33
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 2-Aminohypoxanthine Sulfate

Formula: C₅H₅N₅O
MDL Number: MFCD00213671
CAS Number: 10333-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a08ae43dbeb24bc31170fe5b882e3a91411fbe92&vid=a08ae43dbeb24bc31170fe5b882e3a91411fbe92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR