Guanidinium sulfate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

G0166-25G G0166-500G
TCG0166-025GEA 55.3 CAD
TCG0166-025G TCG0166-500G
Guanidinium sulfate ≥99.0% (by titrimetric analysis)
Guanidinium sulphate

CAS Number: 594-14-9
MDL Number: MFCD00013115
Molecular Formula: CH5N3
Molecular Weight: 216.22
Purity/Analysis Method: >99.0% (T)
Form: Crystal

Formula: C₂H₁₂N₆O₄S
MDL Number: MFCD00013115
CAS Number: 594-14-9
EINECS: 209-826-8

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR