Aminoacetamidine dihydrobromide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Aminoacetamidine dihydrobromide

A1361-1G A1361-5G
TCA1361-001GEA 195.92 CAD
TCA1361-001G TCA1361-5G
Aminoacetamidine dihydrobromide ≥95.0% (by titrimetric analysis)
Aminoacetamidine dihydrobromide

CAS Number: 69816-37-1
MDL Number: MFCD00671486
Molecular Formula: C2H7N3
Molecular Weight: 234.92
Purity/Analysis Method: >95.0% (T)
Form: Crystal

Formula: C₂H₇N₃
MW: 234.92 g/mol
MDL Number: MFCD00671486
CAS Number: 69816-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5c6ab00ae3139c4f432ddc1e5f9e1c0d2024a85&vid=d5c6ab00ae3139c4f432ddc1e5f9e1c0d2024a85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR