Orange II ≥97.0% (by HPLC)
Supplier: TCI America
G0158-25G
G0158-100G
TCG0158-025GEA
56.25
CAD
TCG0158-025G
TCG0158-100G
Orange II ≥97.0% (by HPLC)
Orange II
CAS Number: 633-96-5
MDL Number: MFCD00011657
Molecular Formula: C16H12N2O4S
Molecular Weight: 350.32
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 164
Lambda max.: 484 nm (0.02mol/L AcONH4 sol.)
Product Categories: Bioscience, Stains & Dyes
Other Product Names / Synonyms: Gold Orange, p-(2-Hydroxy-1-naphthylazo)benzenesulfonic Acid Sodium Salt, Orange II, Tropaeolin OOO-2
Formula:
C₁₆H₁₁N₂NaO₄S Melting Pt: 164 °C |
MDL Number:
MFCD00011657 CAS Number: 633-96-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...