Glyoxalbis(2-hydroxyanil) ≥98.0%
Supplier: TCI America
Synonyms:
2,2'-Ethanediylidenedinitrilodiphenol, 2,2'-(Ethane-1,2-diylidenebis(azanylylidene))diphenol, Glyoxalbis(2-hydroxyanil), 1,2-Bis(2-hydroxyphenylimino)ethane
G0153-5G
G0153-25G
TCG0153-5GEA
105.86
CAD
TCG0153-5G
TCG0153-025G
Glyoxalbis(2-hydroxyanil) ≥98.0%
Glyoxalbis(2-hydroxyanil)
CAS Number: 1149-16-2
MDL Number: MFCD00002183
Molecular Formula: C14H12N2O2
Molecular Weight: 240.26
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 205
Formula:
C₁₄H₁₂N₂O₂ MW: 240.26 g/mol Melting Pt: 205 °C |
MDL Number:
MFCD00002183 CAS Number: 1149-16-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...