Glycyl-L-Valine

Supplier: TCI America

G0148-5G G0148-1G
TCG0148-5GEA 469.31 CAD
TCG0148-5G TCG0148-001G
Glycyl-L-Valine
Proteins and Peptides

Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Val-OH


  • Specific rotation [a]20/D: –20° (C=2, H₂O)
  • Solubility in water: Almost transparency
  • EINECS: 217-806-5


Product Categories: Bioscience, Oligopeptides Other Product Names / Synonyms: H-Gly-Val-OH

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17ba7588ffe3d82fa830935c149e70442bcb1cd8&vid=17ba7588ffe3d82fa830935c149e70442bcb1cd8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR