Glycyl-L-tyrosine hydrate
Supplier: TCI America
G0145-5G
G0145-1G
TCG0145-5GEA
425.92
CAD
TCG0145-5G
TCG0145-001G
Glycyl-L-tyrosine hydrate
Proteins and Peptides
CAS Number: 658-79-7
MDL Number: MFCD00008126
Molecular Formula: C11H14N2O4
Molecular Weight: 238.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 47.5 deg (C=1, H2O)
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Tyr-OH
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...