Glycyl-L-Phenylalanine

Supplier: TCI America

G0136-1G G0136-100MG
TCG0136-001GEA 205.4 CAD
TCG0136-001G TCG0136-100MG
Glycyl-L-Phenylalanine
Proteins and Peptides

Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Phe-OH


  • Specific rotation [a]20/D: 41° (C=2, H₂O)
  • Solubility in water: almost transparency
  • EINECS: 222-027-9


Product Categories: Bioscience, Oligopeptides Other Product Names / Synonyms: H-Gly-Phe-OH

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba28ce0b6cc75f3e75ae10115d1c2f5af8820236&vid=ba28ce0b6cc75f3e75ae10115d1c2f5af8820236&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR