(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one ≥98.0%

Supplier: TCI America

Synonyms: 2-azabicyclo[2.2.1]hept-5-en-3-one

A1359-5G A1359-1G A1359-25G
TCA1359-5GEA 41.08 CAD
TCA1359-5G TCA1359-001G TCA1359-25G
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one ≥98.0%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one

CAS Number: 49805-30-3
MDL Number: MFCD00213364
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 57
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Amino-2-cyclopentene-1-carboxylic Acid Lactam

Formula: C₆H₇NO
MW: 109.1259 g/mol
Melting Pt: 57 °C
Flash Pt: 110 °C
MDL Number: MFCD00213364
CAS Number: 49805-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbd73eaae3cc10bca80305b705c2d1d4b82988f5&vid=fbd73eaae3cc10bca80305b705c2d1d4b82988f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR