Glycyl-DL-norleucine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-Gly-DL-Nle-OH

G0133-1G G0133-5G
TCG0133-001GEA 88.48 CAD
TCG0133-001G TCG0133-5G
Glycyl-DL-norleucine ≥98.0% (by titrimetric analysis)
Glycyl-DL-norleucine

CAS Number: 1504-41-2
MDL Number: MFCD00038187
Molecular Formula: C8H16N2O3
Molecular Weight: 188.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 227


Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-DL-Nle-OH

Formula: C₈H₁₆N₂O₃
MW: 188.23 g/mol
Melting Pt: 227 °C
MDL Number: MFCD00038187
CAS Number: 1504-41-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR