Glycyl-D-leucine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

G0130-1G G0130-100MG
TCG0130-001GEA 124.03 CAD
TCG0130-001G TCG0130-100MG
Glycyl-D-leucine ≥99.0% (by titrimetric analysis)
Glycyl-D-leucine

CAS Number: 688-13-1
MDL Number: MFCD00066050
Molecular Formula: C8H16N2O3
Molecular Weight: 188.23
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 234
Specific rotation [a]20/D: 35 deg (C=2, H2O)


Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-D-Leu-OH

Formula: C₈H₁₆N₂O₃
Melting Pt: 234 °C
MDL Number: MFCD00066050
CAS Number: 688-13-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=082d29c0fe4c8574a88f7df0d693ed39c22b4bf9&vid=082d29c0fe4c8574a88f7df0d693ed39c22b4bf9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR