Glycyl-L-Glutamic Acid
Supplier: TCI America
G0123-1G
G0123-100MG
TCG0123-1GEA
213.3
CAD
TCG0123-1G
TCG0123-100MG
Glycyl-L-Glutamic Acid
Proteins and Peptides
Melting point: 158 °C
Specific rotation [a]20/D: –7° (C=5, H₂O)
Water: Max. 8.0%
Purity (nonaqueous titration): Min. 98.0% (calculated on anhydrous substance)
Product categories: Bioscience, Oligopeptides
Other product names/synonyms: H-Gly-Glu-OH
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...