1,3-Adamantanedicarboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

A1358-5G A1358-25G
TCA1358-005GEA 143.78 CAD
TCA1358-005G TCA1358-025G
1,3-Adamantanedicarboxylic acid ≥97.0% (by GC, titration analysis)
1,3-Adamantanedicarboxylic acid

CAS Number: 39269-10-8
MDL Number: MFCD00167790
Molecular Formula: C12H16O4
Molecular Weight: 224.26
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Monomers
Other Product Names / Synonyms: 1,3-Dicarboxyadamantane

Formula: C₁₂H₁₆O₄
MDL Number: MFCD00167790
CAS Number: 39269-10-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c49ab11c4e61c46826dcc597947bd0a1d1b56b19&vid=c49ab11c4e61c46826dcc597947bd0a1d1b56b19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR