Glycy- L-alanine
Supplier: TCI America
G0113-1G
G0113-5G
TCG0113-001GEA
97.96
CAD
TCG0113-001G
TCG0113-5G
Glycy- L-alanine
Proteins and Peptides
CAS Number: 3695-73-6
MDL Number: MFCD00065112
Molecular Formula: C5H10N2O3
Molecular Weight: 146.15
Purity/Analysis Method:
- Specific rotation [a]20/D: –48.0 to –51.0 °C (C=2, H₂O)
- Solubility in water: Almost transparency
- EINECS: 223-019-8
- Product Categories: Bioscience, Oligopeptides
- Other Product Names / Synonyms: H-Gly-Ala-OH
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Ala-OH
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...