di-Sodium β-glycerophosphate pentahydrate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
G0097-25G
G0097-500G
TCG0097-025GEA
196.92
CAD
TCG0097-025G
TCG0097-500G
di-Sodium β-glycerophosphate pentahydrate ≥95.0% (by titrimetric analysis)
di-Sodium β-glycerophosphate pentahydrate
CAS Number: 13408-09-8
MDL Number: MFCD00002135
Molecular Formula: C3H9O6P
Molecular Weight: 216.04
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 102
Other Product Names / Synonyms: beta-Glycerophosphoric Acid Disodium Salt
Formula:
C₃H₇Na₂O₆P·5H₂O Melting Pt: 102 °C |
MDL Number:
MFCD00002135 CAS Number: 13408-09-8 EINECS: 212-464-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...