Triolein 80,0

Supplier: TCI America

Synonyms: Glyceryl trioleate, Glycerine trioleate, Glycerol Trioleate

G0089-100G G0089-25G
TCG0089-100GEA 451.25 CAD
TCG0089-100G TCG0089-025G
Triolein 80,0
Triolein

CAS Number: 122-32-7
MDL Number: MFCD00137563
Molecular Formula: C57H104O6
Molecular Weight: 885.45
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Boiling point (°C): 240
Melting point (°C): -4
Specific Gravity (20/20): 0.92


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Trioleate

Formula: C₅₇H₁₀₄O₆
MW: 885.45 g/mol
Boiling Pt: 240 °C
Melting Pt: –4 °C
Density: 0.92 (20 °C)
MDL Number: MFCD00137563
CAS Number: 122-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70538f39afe91c57be2e28ee55f3464525909c57&vid=70538f39afe91c57be2e28ee55f3464525909c57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR