Monolaurin ≥98.0%

Supplier: TCI America

G0081-25G G0081-5G
TCG0081-025GEA 387.8 CAD
TCG0081-025G TCG0081-5G
Monolaurin ≥98.0%
Monolaurin

CAS Number: 142-18-7
MDL Number: MFCD00037815
Molecular Formula: C15H30O4
Molecular Weight: 274.40
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 186
Melting point (°C): 63


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol alpha-Monolaurate

Formula: C₁₅H₃₀O₄
Boiling Pt: 186 °C
Melting Pt: 63 °C
MDL Number: MFCD00037815
CAS Number: 142-18-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bf5fbdacb89296265381086f273cce0441d95db&vid=3bf5fbdacb89296265381086f273cce0441d95db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR