Disodium glutarate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
G0070-500G
G0070-25G
TCG0070-500GEA
987.5
CAD
TCG0070-500G
TCG0070-025G
Disodium glutarate ≥99.0% (by titrimetric analysis)
Disodium glutarate
CAS Number: 13521-83-0
MDL Number: MFCD00070497
Molecular Formula: C5H8O4
Molecular Weight: 176.08
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Other Product Names / Synonyms: Glutaric Acid Disodium Salt
Formula:
C₅H₈O₄ |
MDL Number:
MFCD00070497 CAS Number: 13521-83-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...