DL-Glutamine ≥98.0%

Supplier: TCI America

Synonyms: H-DL-Gln-OH, 2,5-Diamino-5-oxopentanoic acid

G0062-10G G0062-1G
TCG0062-010GEA 292.62 CAD
TCG0062-010G TCG0062-001G
DL-Glutamine ≥98.0%
DL-Glutamine

CAS Number: 6899-04-3
MDL Number: MFCD00065103
Molecular Formula: C5H10N2O3
Molecular Weight: 146.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Gln-OH

Formula: C₅H₁₀N₂O₃
MW: 146.144 g/mol
MDL Number: MFCD00065103
CAS Number: 6899-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=147b51b7ca8bbf046d7be6266f5f7c0594c9ae31&vid=147b51b7ca8bbf046d7be6266f5f7c0594c9ae31&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR