DL-Glutamic acid 92,0

Supplier: TCI America

G0058-25G G0058-500G
TCG0058-025GEA 79 CAD
TCG0058-025G TCG0058-500G
DL-Glutamic acid 92,0
DL-Glutamic acid

CAS Number: 617-65-2
MDL Number: MFCD00063113
Molecular Formula: C5H9NO4
Molecular Weight: 147.13
Purity/Analysis Method: >92.0% (HPLC)
Form: Crystal
Melting point (°C): 194


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Glu-OH

Formula: C₅H₉NO₄
MW: 147.13 g/mol
Melting Pt: 194 °C
MDL Number: MFCD00063113
CAS Number: 617-65-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8422175d72c3e2fad06593ecc40c894086810a5&vid=c8422175d72c3e2fad06593ecc40c894086810a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR