D-Glucosamic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

G0042-1G G0042-5G
TCG0042-001GEA 263.86 CAD
TCG0042-001G TCG0042-5G
D-Glucosamic acid ≥98.0% (by titrimetric analysis)
D-Glucosamic acid

CAS Number: 3646-68-2
MDL Number: MFCD00037764
Molecular Formula: C6H13NO6
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -15 deg (C=2.5, 1mol/L HCl)


Product Categories: Bioscience, Sugar Acids, Glucosamine, Aminosugars
Other Product Names / Synonyms: D-Glucosaminic Acid

Formula: C₆H₁₃NO₆
MDL Number: MFCD00037764
CAS Number: 3646-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6b36de0516aa59660a2dc4e66fa4ce72a79f927&vid=a6b36de0516aa59660a2dc4e66fa4ce72a79f927&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR