Gibberellic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

G0029-100MG G0029-5G G0029-1G
TCG0029-100MGEA 31.6 CAD
TCG0029-100MG TCG0029-5G TCG0029-001G
Gibberellic acid ≥98.0% (by titrimetric analysis)
Gibberellic acid

CAS Number: 77-06-5
MDL Number: MFCD00079329
Molecular Formula: C19H22O6
Molecular Weight: 346.38
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 235
Specific rotation [a]20/D: 81 deg (C=2, MeOH)


Product Categories: Bioscience, Gibberellins, Diterpenes
Other Product Names / Synonyms: Gibberellic Acid

Formula: C₁₉H₂₂O₆
Melting Pt: 235 °C
Density: 1.386 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00079329
CAS Number: 77-06-5
EINECS: 201-001-0
Merck Index: 13,04430
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1008e1006a13807990c6e095eff7012027addcd3&vid=1008e1006a13807990c6e095eff7012027addcd3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR