Galvinoxyl free radical

Supplier: TCI America

G0020-5G G0020-1G
TCG0020-5GEA 1101.26 CAD
TCG0020-5G TCG0020-001G
Galvinoxyl free radical
Galvinoxyl

CAS Number: 2370-18-5
MDL Number: MFCD00001616
Molecular Formula: C29H41O2
Molecular Weight: 421.65
Form: Crystal
Color: Deep Blue
Storage Temperature: 0-10°C

Formula: C₂₉H₄₁O₂
MDL Number: MFCD00001616
CAS Number: 2370-18-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfb19ac468354d1abd72758df1d1599e614614c4&vid=cfb19ac468354d1abd72758df1d1599e614614c4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR