Hexadecyl gallate ≥95.0%

Supplier: TCI America

Synonyms: Gallic Acid Cetyl Ester, Cetyl Gallate, Gallic Acid Hexadecyl Ester

G0013-5G G0013-25G
TCG0013-005GEA 71.1 CAD
TCG0013-005G TCG0013-025G
Hexadecyl gallate ≥95.0%
Hexadecyl gallate

CAS Number: 5026-65-3
MDL Number: MFCD00016428
Molecular Formula: C23H38O5
Molecular Weight: 394.55
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 100


Other Product Names / Synonyms: Cetyl Gallate, Gallic Acid Cetyl Ester, Gallic Acid Hexadecyl Ester

Formula: C₂₃H₃₈O₅
MW: 394.55 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00016428
CAS Number: 5026-65-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ad07285c98898193ae10f5f883530d247d4bdf6&vid=4ad07285c98898193ae10f5f883530d247d4bdf6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR